Assay Details
Gene Symbol: STIMATE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130713.2 | XP_014986199.1 | 457 | 112 | ||
GenBank mRNA | JU327281.1 | - | 191 | 112 | ||
JV044746.1 | - | 371 | 112 | |||
JV636138.1 | - | 364 | 112 | |||
JV714366.1 | - | 192 | 112 |

More Information
Set Membership: |
![]() ![]() |
