Assay Details
| Gene Symbol: NAPEPLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086505.4 | XP_001086505.3 | 877 | 97 | ||
| XM_015134570.2 | XP_014990056.1 | 716 | 97 | |||
| XM_015134571.2 | XP_014990057.1 | 384 | 97 | |||
| XM_015134572.2 | XP_014990058.1 | 399 | 97 | |||
| XM_015134573.2 | XP_014990059.1 | 491 | 97 | |||
| XM_015134574.2 | XP_014990060.1 | 417 | 97 | |||
| XM_028846142.1 | XP_028701975.1 | 316 | 97 | |||
| GenBank mRNA | JU321104.1 | - | 608 | 97 | ||
| JU471371.1 | - | 363 | 97 | |||
More Information
Set Membership: |
|

NAPEPLD
Genomic Map