Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261478.1 | NP_001248407.1 | 1244 | 68 | ||
| XM_015121080.2 | XP_014976566.1 | 1657 | 68 | |||
| XM_015121085.2 | XP_014976571.1 | 1752 | 68 | |||
| XM_015121086.2 | XP_014976572.1 | 1390 | 68 | |||
| XM_015121094.2 | XP_014976580.1 | 1160 | 68 | |||
| XM_015121101.2 | XP_014976587.1 | 1282 | 68 | |||
| XM_015121103.2 | XP_014976589.1 | 1562 | 68 | |||
| GenBank mRNA | JU320869.1 | - | 1244 | 68 | ||
| JV466999.1 | - | 1408 | 68 | |||
| JV489310.1 | - | 1407 | 68 | |||
More Information
Set Membership: |
|

BRINP3
Genomic Map