Assay Details
| Gene Symbol: ZSWIM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111415.4 | XP_001111415.4 | 880 | 149 | ||
| XM_015124442.2 | XP_014979928.2 | 648 | 149 | |||
| XM_028838575.1 | XP_028694408.1 | 1097 | 149 | |||
More Information
Set Membership: |
|

ZSWIM9
Genomic Map