Assay Details
| Gene Symbol: CERS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266123.2 | NP_001253052.1 | 959 | 71 | ||
| XM_015110252.2 | XP_014965738.1 | 1045 | 71 | |||
| GenBank mRNA | JU475026.1 | - | 961 | 71 | ||
More Information
Set Membership: |
|

CERS6
Genomic Map