Assay Details
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085216.4 | XP_001085216.3 | 750 | 105 | ||
XM_015114091.2 | XP_014969577.2 | 590 | 105 | |||
XM_028833379.1 | XP_028689212.1 | 643 | 105 | |||
XM_028833380.1 | XP_028689213.1 | 698 | 105 | |||
XM_028833381.1 | XP_028689214.1 | 603 | 105 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
