Assay Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261671.1 | NP_001248600.1 | 311 | 78 | ||
XM_015123574.2 | XP_014979060.1 | 618 | 78 | |||
XM_015123575.2 | XP_014979061.1 | 420 | 78 | |||
XM_015123578.2 | XP_014979064.1 | 377 | 78 | |||
XM_015123579.2 | XP_014979065.1 | 525 | 78 | |||
XM_028838460.1 | XP_028694293.1 | 623 | 78 | |||
GenBank mRNA | JU471728.1 | - | 311 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
