Assay Details
| Gene Symbol: SCIMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118383.2 | XP_014973869.1 | 805 | 133 | ||
| GenBank mRNA | JU486300.1 | - | 75 | 133 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCIMP| Gene Symbol: SCIMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118383.2 | XP_014973869.1 | 805 | 133 | ||
| GenBank mRNA | JU486300.1 | - | 75 | 133 | ||
Set Membership: |
|