Assay Details
| Gene Symbol: TMEM207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093355.4 | XP_001093355.2 | 408 | 83 | ||
| XM_028843564.1 | XP_028699397.1 | 379 | 83 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM207| Gene Symbol: TMEM207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093355.4 | XP_001093355.2 | 408 | 83 | ||
| XM_028843564.1 | XP_028699397.1 | 379 | 83 | |||
Set Membership: |
|