Assay Details
| Gene Symbol: ALKAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194437.2 | NP_001181366.1 | 329 | 113 | ||
| XM_015145342.2 | XP_015000828.1 | 1576 | 113 | |||
More Information
Set Membership: |
|

ALKAL1
Genomic Map