Assay Details
| Gene Symbol: CCSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138795.2 | XP_014994281.2 | 2177 | 135 | ||
| XM_015138796.2 | XP_014994282.2 | 2171 | 135 | |||
| XM_015138798.2 | XP_014994284.2 | 2172 | 135 | |||
| XM_028848627.1 | XP_028704460.1 | 2171 | 135 | |||
| XM_028848628.1 | XP_028704461.1 | 2171 | 135 | |||
More Information
Set Membership: |
|

CCSER1
Genomic Map