Assay Details
| Gene Symbol: SLC16A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086787.4 | XP_001086787.2 | 641 | 90 | ||
| XM_015147652.2 | XP_015003138.1 | 2642 | 90 | |||
| XM_015147653.2 | XP_015003139.1 | 408 | 90 | |||
| XM_028826637.1 | XP_028682470.1 | 357 | 90 | |||
More Information
Set Membership: |
|

SLC16A12
Genomic Map