Assay Details
| Gene Symbol: TAFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261362.1 | NP_001248291.1 | 194 | 68 | ||
| XM_015130403.2 | XP_014985889.1 | 347 | 68 | |||
| XM_028843134.1 | XP_028698967.1 | 389 | 68 | |||
| GenBank mRNA | JU320855.1 | - | 194 | 68 | ||
| JV600935.1 | - | 284 | 68 | |||
More Information
Set Membership: |
|

TAFA1
Genomic Map