Assay Details
| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125460.2 | XP_014980946.1 | 1631 | 79 | ||
| GenBank mRNA | JU496026.1 | - | 1590 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PKMYT1| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125460.2 | XP_014980946.1 | 1631 | 79 | ||
| GenBank mRNA | JU496026.1 | - | 1590 | 79 | ||
Set Membership: |
|