Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112806.2 | XP_014968292.1 | 503 | 68 | ||
| XM_015112808.2 | XP_014968294.1 | 527 | 68 | |||
| GenBank mRNA | JU496348.1 | - | 386 | 68 | ||
| JU496349.1 | - | 386 | 68 | |||
More Information
Set Membership: |
|

CD8B
Genomic Map