Assay Details
| Gene Symbol: ALDH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU327098.1 | - | 1070 | 95 | ||
| JV635826.1 | - | 2150 | 95 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALDH1B1| Gene Symbol: ALDH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU327098.1 | - | 1070 | 95 | ||
| JV635826.1 | - | 2150 | 95 | |||
Set Membership: |
|