Assay Details
| Gene Symbol: CRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094433.4 | XP_001094433.1 | 440 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001094433.4 transcripts.
Set Membership: |
|

CRH
Genomic Map