Assay Details
| Gene Symbol: HTR1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090804.4 | XP_001090804.1 | 2026 | 138 | ||
| XM_028847383.1 | XP_028703216.1 | 2088 | 138 | |||
| XM_028847384.1 | XP_028703217.1 | 2133 | 138 | |||
More Information
Set Membership: |
|

HTR1E
Genomic Map