Assay Details
| Gene Symbol: ETFRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151132.2 | XP_015006618.1 | 247 | 139 | ||
| XM_015151133.2 | XP_015006619.1 | 241 | 143 | |||
| GenBank mRNA | JV602185.1 | - | 215 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015151132.2, XM_015151133.2 transcripts.
Set Membership: |
|

ETFRF1
Genomic Map