Assay Details
| Gene Symbol: ATP5F1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032901.2 | NP_001028073.1 | 245 | 164 | ||
| GenBank mRNA | AY341883.1 | - | 245 | 164 | ||
| JV045620.1 | - | 248 | 164 | |||
| JV470859.1 | - | 159 | 164 | |||
| JV602644.1 | - | 268 | 164 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ATP5F1E
Genomic Map