Assay Details
| Gene Symbol: SP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082704.4 | XP_001082704.1 | 633 | 146 | ||
| GenBank mRNA | JV651057.1 | - | 215 | 146 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SP5| Gene Symbol: SP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082704.4 | XP_001082704.1 | 633 | 146 | ||
| GenBank mRNA | JV651057.1 | - | 215 | 146 | ||
Set Membership: |
|