Assay Details
| Gene Symbol: ITPRIPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098586.4 | XP_001098586.1 | 227 | 126 | ||
| GenBank mRNA | JU481051.1 | - | 88 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001098586.4 transcripts.
Set Membership: |
|

ITPRIPL1
Genomic Map