Assay Details
| Gene Symbol: PTCHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136515.2 | XP_014992001.2 | 2468 | 123 | ||
| XM_028847491.1 | XP_028703324.1 | 2642 | 123 | |||
| XM_028847492.1 | XP_028703325.1 | 2963 | 123 | |||
More Information
Set Membership: |
|

PTCHD4
Genomic Map