Assay Details
| Gene Symbol: LOC114678782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849421.1 | XP_028705254.1 | 1319 | 143 | ||
| Gene Symbol: LOC114678780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849419.1 | XP_028705252.1 | 1135 | 143 | ||
| Gene Symbol: LOC114678781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849420.1 | XP_028705253.1 | 2110 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028849419.1, XM_028849420.1, XM_028849421.1 transcripts.
Set Membership: |
|

LOC114678782
Genomic Map