Assay Details
| Gene Symbol: DCAF4L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138293.2 | XP_014993779.1 | 2159 | 153 | ||
| GenBank mRNA | JV045186.1 | - | 1658 | 153 | ||
| JV649029.1 | - | 1678 | 153 | |||
More Information
Set Membership: |
|

DCAF4L1
Genomic Map