Assay Details
| Gene Symbol: MESP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093605.4 | XP_001093605.2 | 862 | 119 | ||
| GenBank mRNA | JU329065.1 | - | 264 | 119 | ||
| JV707519.1 | - | 292 | 119 | |||
More Information
Set Membership: |
|

MESP2
Genomic Map