Assay Details
| Gene Symbol: PGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084229.4 | XP_001084229.2 | 724 | 153 | ||
| GenBank mRNA | JV578998.1 | - | 670 | 153 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PGP| Gene Symbol: PGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084229.4 | XP_001084229.2 | 724 | 153 | ||
| GenBank mRNA | JV578998.1 | - | 670 | 153 | ||
Set Membership: |
|