Assay Details
| Gene Symbol: SST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194239.1 | NP_001181168.1 | 257 | 86 | ||
| GenBank mRNA | JU336076.1 | - | 240 | 86 | ||
| JV568024.1 | - | 243 | 86 | |||
More Information
Set Membership: |
|

SST
Genomic Map