Assay Details
| Gene Symbol: SSTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150285.2 | XP_015005771.1 | 1021 | 133 | ||
| GenBank mRNA | JV048039.1 | - | 2285 | 133 | ||
| JV636335.1 | - | 521 | 133 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015150285.2 transcripts.
Set Membership: |
|

SSTR3
Genomic Map