Assay Details
| Gene Symbol: CLDN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194564.3 | NP_001181493.1 | 137 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194564.3 transcripts.
Set Membership: |
|

CLDN4
Genomic Map