Assay Details
| Gene Symbol: EFNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838772.1 | XP_028694605.1 | 505 | 142 | ||
| GenBank mRNA | JV053899.1 | - | 379 | 142 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EFNA2| Gene Symbol: EFNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838772.1 | XP_028694605.1 | 505 | 142 | ||
| GenBank mRNA | JV053899.1 | - | 379 | 142 | ||
Set Membership: |
|