Assay Details
| Gene Symbol: ID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265656.1 | NP_001252585.1 | 816 | 95 | ||
| GenBank mRNA | JV566450.1 | - | 787 | 95 | ||
| JV601267.1 | - | 849 | 95 | |||
| JV635445.1 | - | 816 | 95 | |||
More Information
Set Membership: |
|

ID4
Genomic Map