Assay Details
| Gene Symbol: ATP6V0C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125387.2 | XP_014980873.1 | 992 | 95 | ||
| GenBank mRNA | JU321313.1 | - | 741 | 95 | ||
| JU476004.1 | - | 672 | 95 | |||
| JV047377.1 | - | 705 | 95 | |||
| JV669619.1 | - | 744 | 95 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ATP6V0C
Genomic Map