Assay Details
| Gene Symbol: FPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116431.4 | XP_001116431.1 | 1113 | 163 | ||
| GenBank mRNA | JU325088.1 | - | 188 | 163 | ||
| JV710963.1 | - | 481 | 163 | |||
More Information
Set Membership: |
|

FPR1
Genomic Map