Assay Details
| Gene Symbol: KCNJ10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257876.1 | NP_001244805.1 | 149 | 71 | ||
| GenBank mRNA | JU320110.1 | - | 223 | 71 | ||
| JU320111.1 | - | 223 | 71 | |||
| JU320112.1 | - | 223 | 71 | |||
| JU475516.1 | - | 149 | 71 | |||
| JU475517.1 | - | 149 | 71 | |||
| JU475518.1 | - | 149 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257876.1 transcripts.
Set Membership: |
|

KCNJ10
Genomic Map