Assay Details
| Gene Symbol: KCNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168626.2 | NP_001162097.1 | 1522 | 109 | ||
| XM_015148832.2 | XP_015004318.2 | 1630 | 109 | |||
| GenBank mRNA | JU327110.1 | - | 120 | 109 | ||
| JU327111.1 | - | 120 | 109 | |||
| JV506086.1 | - | 151 | 109 | |||
| JV565114.1 | - | 1128 | 109 | |||
| JV581208.1 | - | 1128 | 109 | |||
More Information
Set Membership: |
|

KCNS1
Genomic Map