Assay Details
| Gene Symbol: SRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261094.2 | NP_001248023.1 | 245 | 157 | ||
| GenBank mRNA | JU335343.1 | - | 276 | 157 | ||
| JU476295.1 | - | 296 | 157 | |||
| JV047576.1 | - | 247 | 157 | |||
| JV718293.1 | - | 285 | 157 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SRGN
Genomic Map