Assay Details
| Gene Symbol: CCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032874.1 | NP_001028046.1 | 184 | 102 | ||
| GenBank mRNA | AF449270.1 | - | 184 | 102 | ||
| AY049019.1 | - | 184 | 102 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCL11
Genomic Map