Assay Details
| Gene Symbol: TBCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136361.2 | XP_014991847.2 | 1253 | 99 | ||
| GenBank mRNA | JU325542.1 | - | 979 | 99 | ||
| JV637559.1 | - | 1132 | 99 | |||
| JV667250.1 | - | 1129 | 99 | |||
| JV722066.1 | - | 1165 | 99 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TBCC
Genomic Map