Assay Details
| Gene Symbol: TMEM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266370.1 | NP_001253299.1 | 87 | 95 | ||
| GenBank mRNA | JU477662.1 | - | 77 | 95 | ||
| JV636438.1 | - | 87 | 95 | |||
More Information
Set Membership: |
|

TMEM11
Genomic Map