Assay Details
| Gene Symbol: RBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194599.2 | NP_001181528.1 | 534 | 73 | ||
| XM_028843394.1 | XP_028699227.1 | 467 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RBP2
Genomic Map