Assay Details
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106464.4 | XP_001106464.2 | 1437 | 147 | ||
| XM_015147419.2 | XP_015002905.1 | 2486 | 147 | |||
| GenBank mRNA | JU337190.1 | - | 426 | 147 | ||
| JV048760.1 | - | 556 | 147 | |||
| JV666088.1 | - | 591 | 147 | |||
More Information
Set Membership: |
|

CHST3
Genomic Map