Assay Details
| Gene Symbol: NAP1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265658.1 | NP_001252587.1 | 45 | 62 | ||
| GenBank mRNA | JV667400.1 | - | 45 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265658.1 transcripts.
Set Membership: |
|

NAP1L3
Genomic Map