Assay Details
| Gene Symbol: SIAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194383.3 | NP_001181312.1 | 1051 | 60 | ||
| GenBank mRNA | JU328359.1 | - | 251 | 60 | ||
| JV635761.1 | - | 750 | 60 | |||
| JV721290.1 | - | 242 | 60 | |||
More Information
Set Membership: |
|

SIAH2
Genomic Map