Assay Details
| Gene Symbol: CCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084047.4 | XP_001084047.1 | 119 | 103 | ||
| GenBank mRNA | JU480021.1 | - | 124 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001084047.4 transcripts.
Set Membership: |
|

CCR8
Genomic Map