Assay Details
| Gene Symbol: GPR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082031.4 | XP_001082031.1 | 104 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001082031.4 transcripts.
Set Membership: |
|

GPR20
Genomic Map