Assay Details
| Gene Symbol: ZBTB22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136108.2 | XP_014991594.2 | 1309 | 156 | ||
| GenBank mRNA | JV048708.1 | - | 980 | 156 | ||
| JV665681.1 | - | 1057 | 156 | |||
More Information
Set Membership: |
|

ZBTB22
Genomic Map