Assay Details
| Gene Symbol: POU3F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136968.2 | XP_014992454.2 | 2689 | 98 | ||
| GenBank mRNA | JU344856.1 | - | 1958 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POU3F2| Gene Symbol: POU3F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136968.2 | XP_014992454.2 | 2689 | 98 | ||
| GenBank mRNA | JU344856.1 | - | 1958 | 98 | ||
Set Membership: |
|