Assay Details
| Gene Symbol: SFN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258131.1 | NP_001245060.1 | 51 | 69 | ||
| GenBank mRNA | JU472112.1 | - | 51 | 69 | ||
| JV712416.1 | - | 53 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001258131.1 transcripts.
Set Membership: |
|

SFN
Genomic Map