Assay Details
| Gene Symbol: NFE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109546.3 | XP_001109546.1 | 450 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001109546.3 transcripts.
Set Membership: |
|

NFE2
Genomic Map